siRNA Details
| Gene Symbol: Znrd1as | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029602.1 | NP_083878.1 | 3 | 223 | ||
| GenBank mRNA | AK006236.1 | - | 3 | 216 | ||
| AK161503.1 | - | 3 | 218 | |||
| BC027007.1 | - | 3 | 223 | 5365474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znrd1as| Gene Symbol: Znrd1as | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029602.1 | NP_083878.1 | 3 | 223 | ||
| GenBank mRNA | AK006236.1 | - | 3 | 216 | ||
| AK161503.1 | - | 3 | 218 | |||
| BC027007.1 | - | 3 | 223 | 5365474 | ||