siRNA Details
| Gene Symbol: 3110002H16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029623.2 | NP_083899.1 | 7 | 680 | ||
| GenBank mRNA | AK010309.1 | - | 7 | 674 | ||
| AK013979.1 | - | 2 | 106 | |||
| AK144623.1 | - | 7 | 681 | |||
| AK159691.1 | - | 7 | 675 | |||
| BC012285.1 | - | 7 | 658 | |||
| BC021846.1 | - | 7 | 663 | 5027035 | ||

3110002H16Rik
Genomic Map