siRNA Details
| Gene Symbol: Lmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029624.4 | NP_083900.3 | 2, 3 | 510 | ||
| GenBank mRNA | AK010312.1 | - | 2, 3 | 481 | ||
| AK035119.1 | - | 2, 3 | 498 | |||
| AK053241.1 | - | 2, 3 | 487 | |||
| AK142235.1 | - | 2, 3 | 486 | |||
| AK154622.1 | - | 2, 3 | 507 | |||
| AK169983.1 | - | 2, 3 | 508 | |||
| AK172374.1 | - | 2, 3 | 489 | |||
| BC020104.1 | - | 2, 3 | 517 | 3963699 | ||

Lmf1
Genomic Map