siRNA Details
| Gene Symbol: Lmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029624.4 | NP_083900.3 | 8 | 1163 | ||
| GenBank mRNA | AK010312.1 | - | 8 | 1134 | ||
| AK041659.1 | - | 6 | 830 | |||
| AK154622.1 | - | 8 | 1160 | |||
| AK177501.1 | - | 3 | 296 | |||
| BC020104.1 | - | 8 | 1170 | 3963699 | ||

Lmf1
Genomic Map