siRNA Details
| Gene Symbol: Styxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289554.1 | NP_001276483.1 | 7 | 785 | ||
| NM_001289555.1 | NP_001276484.1 | 7 | 782 | |||
| NM_001289556.1 | NP_001276485.1 | 6 | 626 | |||
| NM_001289557.1 | NP_001276486.1 | 5 | 477 | |||
| NM_029659.4 | NP_083935.3 | 7 | 792 | |||
| GenBank mRNA | AK005599.1 | - | 7 | 783 | ||
| AK005858.1 | - | 6 | 533 | |||
| AK161310.1 | - | 7 | 793 | |||
| BC045148.1 | - | 7 | 722 | 6307739 | ||
| BF471066.1 | - | 5 | 386 | |||

Styxl1
Genomic Map