siRNA Details
| Gene Symbol: Spatc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029661.1 | NP_083937.1 | 1 | 249 | ||
| GenBank mRNA | AK006410.1 | - | 1 | 249 | ||
| BC049686.1 | - | 1 | 240 | |||
| BC147585.1 | - | 1 | 229 | 9056575 | ||
| BC147614.1 | - | 1 | 229 | 9056604 | ||

Spatc1l
Genomic Map