siRNA Details
| Gene Symbol: 1700123K08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029693.2 | NP_083969.1 | 5 | 702 | ||
| GenBank mRNA | AK007254.1 | - | 5 | 703 | ||
| BC147525.1 | - | 5 | 690 | 9056517 | ||
| BC147536.1 | - | 5 | 690 | 9056527 | ||

1700123K08Rik
Genomic Map