siRNA Details
| Gene Symbol: 1700123K08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029693.2 | NP_083969.1 | 6 | 1026 | ||
| GenBank mRNA | AK007254.1 | - | 6 | 1027 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700123K08Rik| Gene Symbol: 1700123K08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029693.2 | NP_083969.1 | 6 | 1026 | ||
| GenBank mRNA | AK007254.1 | - | 6 | 1027 | ||