siRNA Details
| Gene Symbol: Cluap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029738.2 | NP_084014.1 | 4 | 386 | ||
| GenBank mRNA | AK011846.1 | - | 5 | 414 | ||
| AK134648.1 | - | 4 | 353 | |||
| AK166050.1 | - | 4 | 467 | |||
| BC024910.1 | - | 4 | 386 | 3603454 | ||

Cluap1
Genomic Map