siRNA Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029746.3 | NP_084022.2 | 9 | 1028 | ||
| GenBank mRNA | AK089920.1 | - | 9 | 1012 | ||
| AK157348.1 | - | 9 | 990 | |||
| AK169298.1 | - | 9 | 998 | |||
| AK171726.1 | - | 9 | 947 | |||
| BC011435.1 | - | 9 | 968 | 4010102 | ||
| CO423795.1 | - | 6 | 585 | |||
| CX205121.1 | - | 3 | 237 | |||

Cog2
Genomic Map