siRNA Details
| Gene Symbol: Cog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029746.3 | NP_084022.2 | 18 | 2785 | ||
| GenBank mRNA | AK004441.1 | - | 6 | 1262 | ||
| AK089920.1 | - | 18 | 2772 | |||
| AK169298.1 | - | 18 | 2755 | |||
| AK171726.1 | - | 18 | 2704 | |||
| BC011435.1 | - | 18 | 2725 | 4010102 | ||

Cog2
Genomic Map