siRNA Details
| Gene Symbol: Sdccag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029756.3 | NP_084032.1 | 2 | 420 | ||
| GenBank mRNA | AK012392.1 | - | 2 | 208 | ||
| AK017691.1 | - | 2 | 203 | |||
| AK051683.1 | - | 2 | 223 | |||
| AK132316.1 | - | 2 | 216 | |||
| AK133622.1 | - | 2 | 164 | |||
| AK136921.1 | - | 2 | 243 | |||
| BC016244.1 | - | 2 | 200 | 4507575 | ||
| BC025138.1 | - | 2 | 190 | 4483636 | ||
| CJ072722.1 | - | 2 | 272 | |||

Sdccag8
Genomic Map