siRNA Details
| Gene Symbol: Sdccag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029756.3 | NP_084032.1 | 4 | 688 | ||
| GenBank mRNA | AK012392.1 | - | 4 | 476 | ||
| AK017691.1 | - | 4 | 471 | |||
| AK051683.1 | - | 4 | 491 | |||
| AK132316.1 | - | 4 | 484 | |||
| AK133622.1 | - | 4 | 432 | |||
| AK136921.1 | - | 4 | 511 | |||
| BC016244.1 | - | 4 | 468 | 4507575 | ||

Sdccag8
Genomic Map