siRNA Details
| Gene Symbol: Fam49a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146119.1 | NP_001139591.1 | 12 | 2249 | ||
| NM_029758.4 | NP_084034.2 | 13 | 2322 | |||
| GenBank mRNA | AK036232.1 | - | 1 | 315 | ||
| BC052465.1 | - | 12 | 2230 | 6834768 | ||

Fam49a
Genomic Map