siRNA Details
| Gene Symbol: Mtfr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256112.1 | NP_001243041.1 | 5 | 716 | ||
| NM_029759.4 | NP_084035.1 | 5 | 667 | |||
| GenBank mRNA | AK010825.1 | - | 5 | 708 | ||
| AK080126.1 | - | 4 | 3036 | |||
| AK145362.1 | - | 5 | 708 | |||
| AK146416.1 | - | 4 | 543 | |||
| AK164004.1 | - | 4 | 543 | |||
| AK169047.1 | - | 5 | 575 | |||
| AK171703.1 | - | 5 | 708 | |||
| BC026808.1 | - | 4 | 539 | 4159289 | ||

Mtfr1l
Genomic Map