siRNA Details
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB095735.1 | - | 6 | 631 | ||
| AB095736.1 | - | 6 | 631 | |||
| AK012919.1 | - | 6 | 632 | |||
| AK054412.1 | - | 6 | 621 | |||
| AK133177.1 | - | 6 | 630 | |||
| AK154594.1 | - | 7 | 671 | |||
| AK161401.1 | - | 3 | 232 | |||
| AK164249.1 | - | 6 | 631 | |||
| AK164999.1 | - | 6 | 630 | |||
| AK166351.1 | - | 6 | 639 | |||
| AK169601.1 | - | 6 | 630 | |||
| BC060208.1 | - | 6 | 590 | |||

Dtl
Genomic Map