siRNA Details
| Gene Symbol: Mettl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029790.3 | NP_084066.2 | 3 | 450 | ||
| GenBank mRNA | AK002682.1 | - | 3 | 451 | ||
| BC089533.1 | - | 4 | 577 | |||
| BC106186.1 | - | 3 | 422 | 30244889 | ||
| BC115926.1 | - | 3 | 409 | 40105262 | ||
| BC117549.1 | - | 3 | 409 | 40105259 | ||

Mettl15
Genomic Map