siRNA Details
| Gene Symbol: RP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006269.1 | NP_006260.1 | 2 | 699 | ||
| XM_017013721.1 | XP_016869210.1 | 7 | 1862 | |||
| XM_017013722.1 | XP_016869211.1 | 2 | 715 | |||
| GenBank mRNA | AF141021.1 | - | 2 | 699 | ||
| AF143222.1 | - | 2 | 649 | |||
| AF146592.1 | - | 2 | 647 | |||
| AK098176.1 | - | 2, 3 | 337 | |||
| AK302348.1 | - | 1 | 83 | |||
| AK304443.1 | - | 3 | 244 | |||
| AK316511.1 | - | 3 | 678 | |||

RP1
Genomic Map