siRNA Details
| Gene Symbol: Golga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290649.1 | NP_001277578.1 | 22 | 2721 | ||
| NM_029793.2 | NP_084069.1 | 23 | 2796 | |||
| GenBank mRNA | AK163007.1 | - | 3 | 369 | ||
| BC037037.1 | - | 18 | 2132 | |||
| BC043090.1 | - | 23 | 2661 | 6418353 | ||
| BC053068.1 | - | 22 | 2649 | 6405595 | ||
| BC080733.1 | - | 22 | 2721 | 30110235 | ||

Golga1
Genomic Map