siRNA Details
| Gene Symbol: 2510039O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029841.3 | NP_084117.2 | 3 | 2339 | ||
| GenBank mRNA | AK011059.1 | - | 3 | 2339 | ||
| AK173332.1 | - | 3 | 3333 | |||
| BC012878.1 | - | 3 | 2312 | 4190292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2510039O18Rik| Gene Symbol: 2510039O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029841.3 | NP_084117.2 | 3 | 2339 | ||
| GenBank mRNA | AK011059.1 | - | 3 | 2339 | ||
| AK173332.1 | - | 3 | 3333 | |||
| BC012878.1 | - | 3 | 2312 | 4190292 | ||