siRNA Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029852.2 | NP_084128.2 | 3, 4 | 684 | ||
| GenBank mRNA | AK015007.1 | - | 3, 4 | 821 | ||
| AK141627.1 | - | 3, 4 | 805 | |||
| BC028960.1 | - | 2, 3 | 385 | |||
| BC043468.1 | - | 2, 3 | 298 | |||
| BC070464.1 | - | 3, 4 | 684 | 30608497 | ||

Cep83
Genomic Map