siRNA Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029852.2 | NP_084128.2 | 15 | 2201 | ||
| GenBank mRNA | AK015007.1 | - | 15 | 2338 | ||
| AK151291.1 | - | 9 | 1050 | |||
| AK152465.1 | - | 9 | 1050 | |||
| AK152801.1 | - | 9 | 1050 | |||
| AK153515.1 | - | 9 | 1050 | |||
| BC028960.1 | - | 14 | 1901 | |||
| BC043468.1 | - | 14 | 1815 | |||
| BC070464.1 | - | 15 | 2201 | 30608497 | ||

Cep83
Genomic Map