siRNA Details
| Gene Symbol: Ankrd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168403.1 | NP_001161875.1 | 12 | 2745 | ||
| NM_029898.3 | NP_084174.2 | 11 | 2639 | |||
| GenBank mRNA | AK021066.1 | - | 3 | 790 | ||
| AK045480.1 | - | 11 | 2273 | |||

Ankrd55
Genomic Map