siRNA Details
| Gene Symbol: 9530003J23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029906.3 | NP_084182.2 | 4 | 920 | ||
| GenBank mRNA | AK020538.1 | - | 4 | 846 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530003J23Rik| Gene Symbol: 9530003J23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029906.3 | NP_084182.2 | 4 | 920 | ||
| GenBank mRNA | AK020538.1 | - | 4 | 846 | ||