siRNA Details
| Gene Symbol: Tmem116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161626.1 | NP_001155098.1 | 10 | 989 | ||
| NM_001161627.1 | NP_001155099.1 | 11 | 1094 | |||
| NM_029912.2 | NP_084188.1 | 10 | 1027 | |||
| GenBank mRNA | AK021099.1 | - | 10 | 1033 | ||
| BC019151.1 | - | 10 | 984 | 5064772 | ||

Tmem116
Genomic Map