siRNA Details
| Gene Symbol: Lrrc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029985.2 | NP_084261.1 | 2 | 487 | ||
| GenBank mRNA | AK020847.1 | - | 2 | 293 | ||
| AK143870.1 | - | 1 | 261 | |||
| BC027203.1 | - | 2 | 419 | 3710091 | ||
| BC031894.1 | - | 2 | 391 | 5362699 | ||

Lrrc42
Genomic Map