siRNA Details
| Gene Symbol: Armcx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166377.1 | NP_001159849.1 | 6 | 1748 | ||
| NM_001166378.1 | NP_001159850.1 | 4 | 1447 | |||
| NM_001166379.1 | NP_001159851.1 | 4 | 1563 | |||
| NM_001166380.1 | NP_001159852.1 | 5 | 1539 | |||
| NM_030066.3 | NP_084342.1 | 6 | 1632 | |||
| GenBank mRNA | AK019405.1 | - | 6 | 1559 | ||
| AK147002.1 | - | 6 | 1699 | |||
| BC021410.1 | - | 5 | 1467 | 5007058 | ||
| BC026488.1 | - | 4 | 1531 | 4504219 | ||
| BC068228.1 | - | 6 | 1581 | 6413546 | ||

Armcx1
Genomic Map