siRNA Details
| Gene Symbol: Ndufv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030087.2 | NP_084363.2 | 3 | 1089 | ||
| GenBank mRNA | AK154191.1 | - | 3 | 1022 | ||
| AK154477.1 | - | 3 | 1068 | |||
| BC013640.1 | - | 3 | 1049 | 4188234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufv3| Gene Symbol: Ndufv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030087.2 | NP_084363.2 | 3 | 1089 | ||
| GenBank mRNA | AK154191.1 | - | 3 | 1022 | ||
| AK154477.1 | - | 3 | 1068 | |||
| BC013640.1 | - | 3 | 1049 | 4188234 | ||