siRNA Details
| Gene Symbol: Ndufv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083891.1 | NP_001077360.1 | 3 | 323 | ||
| NM_030087.2 | NP_084363.2 | 4 | 1415 | |||
| GenBank mRNA | AK077433.1 | - | 3 | 264 | ||
| AK154191.1 | - | 4 | 1348 | |||
| AK154477.1 | - | 4 | 1394 | |||
| AK201972.1 | - | 1 | 67 | |||
| BC013640.1 | - | 4 | 1375 | 4188234 | ||
| BU555486.1 | - | 3 | 326 | |||

Ndufv3
Genomic Map