siRNA Details
| Gene Symbol: Arhgap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030113.2 | NP_084389.2 | 6 | 660 | ||
| GenBank mRNA | AF297029.1 | - | 6 | 564 | ||
| AF297030.1 | - | 6 | 623 | |||
| AY179965.1 | - | 6 | 536 | |||
| AY179967.1 | - | 6 | 536 | |||
| CV783737.1 | - | 1 | 38 | |||

Arhgap10
Genomic Map