siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 8, 9 | 757 | ||
| GenBank mRNA | AY102701.1 | - | 8, 9 | 679 | ||
| BC027201.1 | - | 8, 9 | 693 | |||
| BC043701.1 | - | 7, 8 | 745 | |||
| BC056435.1 | - | 8, 9 | 666 | 5707801 | ||

Naa35
Genomic Map