siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 17 | 1609 | ||
| GenBank mRNA | AK050805.1 | - | 7 | 944 | ||
| AK177551.1 | - | 4 | 271 | |||
| AY102701.1 | - | 17 | 1531 | |||
| BC056435.1 | - | 17 | 1518 | 5707801 | ||

Naa35
Genomic Map