siRNA Details
| Gene Symbol: Fam122b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166365.2 | NP_001159837.2 | 9 | 1773 | ||
| NM_030167.4 | NP_084443.1 | 9 | 1776 | |||
| GenBank mRNA | AK019480.1 | - | 9 | 1003 | ||
| BC066167.1 | - | 9 | 988 | 30433483 | ||

Fam122b
Genomic Map