siRNA Details
| Gene Symbol: Trmt44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030208.3 | NP_084484.2 | 2 | 597 | ||
| GenBank mRNA | AK019101.1 | - | 2 | 588 | ||
| AK046957.1 | - | 2 | 599 | |||
| AK132402.1 | - | 2 | 598 | |||
| BC152332.1 | - | 2 | 592 | 40141614 | ||

Trmt44
Genomic Map