siRNA Details
| Gene Symbol: Fbxo34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146085.1 | NP_001139557.1 | 4 | 607 | ||
| NM_001146086.1 | NP_001139558.1 | 3 | 530 | |||
| NM_030236.3 | NP_084512.2 | 2 | 420 | |||
| GenBank mRNA | AK013711.1 | - | 6 | 969 | ||
| AK020018.1 | - | 2 | 421 | |||
| AK171630.1 | - | 7 | 955 | |||
| BC048149.1 | - | 4 | 641 | 5706344 | ||
| BC049155.1 | - | 3 | 472 | |||

Fbxo34
Genomic Map