siRNA Details
| Gene Symbol: Parp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030253.3 | NP_084529.1 | 4 | 1383 | ||
| GenBank mRNA | AK037903.1 | - | 4 | 950 | ||
| AK050032.1 | - | 4 | 827 | |||
| BC003281.1 | - | 4 | 825 | 3581841 | ||
| BC010312.1 | - | 3 | 808 | |||
| BC070466.1 | - | 4 | 1383 | 30615225 | ||

Parp9
Genomic Map