siRNA Details
| Gene Symbol: Gpr146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038703.2 | NP_001033792.1 | 2 | 922 | ||
| NM_030258.4 | NP_084534.2 | 2 | 847 | |||
| GenBank mRNA | AK046512.1 | - | 2 | 870 | ||
| AK050034.1 | - | 2 | 816 | |||
| AK087100.1 | - | 2 | 816 | |||
| AK089232.1 | - | 2 | 870 | |||
| AK137454.1 | - | 2 | 924 | |||
| AK154478.1 | - | 2 | 876 | |||
| AK154708.1 | - | 2 | 868 | |||
| AK155212.1 | - | 2 | 815 | |||
| AK155280.1 | - | 2 | 936 | |||
| AK155876.1 | - | 2 | 850 | |||
| BC003323.1 | - | 2 | 818 | 3156060 | ||
More Information
Set Membership: |
|

Gpr146
Genomic Map