siRNA Details
| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030259.1 | NP_084535.1 | 4 | 743 | ||
| GenBank mRNA | AK133760.1 | - | 4 | 875 | ||
| AK150123.1 | - | 4 | 847 | |||
| AK150252.1 | - | 4 | 846 | |||
| BC003324.1 | - | 4 | 743 | 3156139 | ||

Rilpl2
Genomic Map