siRNA Details
| Gene Symbol: Pbx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024954.1 | NP_001020125.1 | 8 | 1329 | ||
| GenBank mRNA | AI605695.1 | - | 303 | |||
| AJ300184.1 | - | 5 | 762 | |||
| AK010506.1 | - | 5 | 808 | |||
| AK038786.1 | - | 933 | ||||
| AK076968.1 | - | 6 | 953 | |||
| BC032875.1 | - | 7 | 1025 | 1263363 | ||

Pbx4
Genomic Map