siRNA Details
| Gene Symbol: BC004004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030561.3 | NP_085038.3 | 9 | 2355 | ||
| GenBank mRNA | AK030596.1 | - | 7 | 1925 | ||
| BB027102.2 | - | 1 | 254 | |||
| BC058575.1 | - | 9 | 2164 | 6488611 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC004004| Gene Symbol: BC004004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030561.3 | NP_085038.3 | 9 | 2355 | ||
| GenBank mRNA | AK030596.1 | - | 7 | 1925 | ||
| BB027102.2 | - | 1 | 254 | |||
| BC058575.1 | - | 9 | 2164 | 6488611 | ||