siRNA Details
| Gene Symbol: Fam20c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030565.6 | NP_085042.2 | 10 | 2564 | ||
| GenBank mRNA | AK154810.1 | - | 10 | 2560 | ||
| AK180985.1 | - | 1 | 184 | |||
| AK220286.1 | - | 10 | 2080 | |||
| AY778962.1 | - | 10 | 2545 | |||
| BC004044.1 | - | 7 | 1173 | |||
| BC025814.1 | - | 10 | 2515 | |||
| BC025826.1 | - | 10 | 2545 | |||

Fam20c
Genomic Map