siRNA Details
| Gene Symbol: Sacm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030692.4 | NP_109617.1 | 3 | 182 | ||
| GenBank mRNA | AJ245720.1 | - | 3 | 312 | ||
| AJ297526.1 | - | 3 | 183 | |||
| AK089174.1 | - | 3 | 266 | |||
| AK122390.1 | - | 3 | 247 | |||
| AK154759.1 | - | 3 | 283 | |||
| AK161111.1 | - | 3 | 216 | |||
| AK171080.1 | - | 3 | 266 | |||
| BC117752.1 | - | 3 | 183 | 40090556 | ||
| BC117753.1 | - | 3 | 183 | 40090558 | ||
| EF437383.1 | - | 3 | 231 | |||

Sacm1l
Genomic Map