siRNA Details
| Gene Symbol: Cxcr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030712.4 | NP_109637.3 | 2 | 640 | ||
| GenBank mRNA | AF301018.1 | - | 2 | 643 | ||
| AF305709.1 | - | 2 | 643 | |||
| AK052901.1 | - | 2 | 638 | |||
| AK137703.1 | - | 2 | 619 | |||
| AK153914.1 | - | 2 | 639 | |||
| BC096491.1 | - | 2 | 660 | 30748222 | ||
| JC209414.1 | - | 2 | 640 | |||

Cxcr6
Genomic Map