siRNA Details
| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030732.3 | NP_109657.2 | 7 | 1101 | ||
| GenBank mRNA | AF268195.1 | - | 7 | 882 | ||
| AK029595.1 | - | 7 | 900 | |||
| AK033347.1 | - | 7 | 971 | |||
| AK036064.1 | - | 8 | 1212 | |||
| AK136255.1 | - | 7 | 923 | |||

Tbl1xr1
Genomic Map