siRNA Details
| Gene Symbol: Ift122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167763.1 | NP_001161235.1 | 4 | 376 | ||
| NM_031177.4 | NP_112454.2 | 4 | 376 | |||
| GenBank mRNA | AK031308.1 | - | 4 | 264 | ||
| AK044456.1 | - | 4 | 376 | |||
| AK131115.1 | - | 4 | 303 | |||
| AK167330.1 | - | 4 | 331 | |||
| AK167412.1 | - | 4 | 331 | |||
| AK199875.1 | - | 3 | 205 | |||
| AK206333.1 | - | 3 | 204 | |||
| AK218729.1 | - | 2 | 137 | |||
| BC060621.1 | - | 4 | 324 | |||
| BC066083.1 | - | 4 | 295 | 30355922 | ||
| BC067415.1 | - | 4 | 292 | 30063124 | ||

Ift122
Genomic Map