siRNA Details
| Gene Symbol: Sp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031183.2 | NP_112460.1 | 2 | 1314 | ||
| GenBank mRNA | AJ275988.1 | - | 1 | 543 | ||
| AK137686.1 | - | 2 | 1537 | |||
| AM295492.1 | - | 2 | 1558 | |||
| AY338955.1 | - | 2 | 1314 | |||
| BC138818.1 | - | 1 | 1260 | 8861840 | ||
| BC138822.1 | - | 1 | 1260 | 8861844 | ||

Sp6
Genomic Map