siRNA Details
| Gene Symbol: Tex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199293.1 | NP_001186222.1 | 12 | 1516 | ||
| NM_031386.2 | NP_113563.2 | 12 | 1497 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex14| Gene Symbol: Tex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199293.1 | NP_001186222.1 | 12 | 1516 | ||
| NM_031386.2 | NP_113563.2 | 12 | 1497 | |||