siRNA Details
| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031842.2 | NP_114030.2 | 13 | 2414 | ||
| GenBank mRNA | AK075611.1 | - | 9 | 1749 | ||
| BC026783.1 | - | 13 | 2326 | 4481097 | ||
| BC059921.1 | - | 13 | 2384 | |||
| BC079839.1 | - | 13 | 2360 | 6810462 | ||
| CX221058.1 | - | 1 | 15 | |||
| M25773.1 | - | 13 | 2363 | |||
| U66620.1 | - | 13 | 2365 | |||

Smarcd1
Genomic Map