siRNA Details
| Gene Symbol: Nedd4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114386.1 | NP_001107858.1 | 20 | 2097 | ||
| NM_031881.2 | NP_114087.2 | 19 | 2146 | |||
| GenBank mRNA | AF277232.1 | - | 19 | 1779 | ||
| AK004969.1 | - | 19 | 1839 | |||
| AK031842.1 | - | 18 | 1767 | |||
| AK042621.1 | - | 19 | 1720 | |||
| AK163497.1 | - | 17 | 1567 | |||
| AK192598.1 | - | 1 | 37 | |||
| BC039746.1 | - | 19 | 1790 | 3157137 | ||
| BC071210.1 | - | 19 | 1767 | 4972426 | ||
| BC082281.1 | - | 16 | 1504 | |||

Nedd4l
Genomic Map