siRNA Details
| Gene Symbol: Tmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033759.2 | NP_001028931.1 | 8 | 2197 | ||
| NM_031997.4 | NP_114386.3 | 9 | 2279 | |||
| GenBank mRNA | AK129352.1 | - | 3 | 498 | ||
| BC076570.1 | - | 8 | 1803 | 6830791 | ||
| BC089353.1 | - | 8 | 2179 | 6410437 | ||
| BQ895808.1 | - | 499 | ||||
| CA752446.1 | - | 1 | 137 | |||

Tmem2
Genomic Map